Key Documents
Z301086
Quickfit® expansion adapter
top female joint: ST/NS 14/23, bottom male joint: ST/NS 10/19
Sinonimo/i:
XA10
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
female joint: ST/NS top14/23
male joint: ST/NS bottom10/19
Produttore/marchio commerciale
Quickfit XA 10
Disponibilità
not available in USA and Canada
not available in USA and Canada
Descrizione generale
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)