Z278769
Bytac® surface protection laminate
PTFE, size 25 in. × 15 ft
Sinonimo/i:
bench protection, benchtop laminate, worksurface protection
About This Item
Codice UNSPSC:
41122300
NACRES:
NB.35
Prodotti consigliati
Materiali
PTFE
vinyl backing
Confezionamento
pack of 1 ea
Misura
25 in. × 15 ft
Categorie correlate
Descrizione generale
Add the benefits of FEP to any flat surface. Bytac protects surfaces from absorption of toxic and radioactive materials. The laminate consists of FEP resin supported on a backing of vinyl or aluminum. The backing has a pressure sensitive acrylic adhesive that adheres to most surfaces. Vinyl backing is recommended for applications up to 200 °F (93 °C). Aluminum backing is for high-temperature applications and can be used up to 360 °F (182 °C).
- non-stick; easy to clean
- chemically inert
Note legali
Bytac is a registered trademark of Saint-Gobain Performance Plastics Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z278769-1EA | 4061833578544 |