Z268747
Scienceware® Safe-Lab™ PTFE stirrer bearing
joint: ST/NS 24/40
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.72
Prodotti consigliati
Materiali
Viton® O-ring
Giunto
joint: ST/NS24/40
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F20320-1661
Categorie correlate
Descrizione generale
Stopper-Loc® nut allows stirrer bearing to be locked to outer ground-glass joint. Complete with Stopper-Loc nut and two PTFE inserts. For 10mm diam. polished glass or PTFE stirring shafts.
Note legali
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Stopper-Loc is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z268747-1EA | 4061832262925 |