Documenti fondamentali
Z265950
Sigma-Aldrich
Solid-glass beads
soda-lime glass, diam. 6 mm
About This Item
Codice UNSPSC:
41102422
NACRES:
NB.43
Materiali
soda-lime glass
Diametro
6 mm
Descrizione generale
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Glass beads 5 mm
Supelco
1.04017
Glass beads 5 mm
Solid-glass beads borosilicate, diam. 1 mm
Z273619
Solid-glass beads
Solid-glass beads borosilicate, diam. 2 mm
Z273627
Solid-glass beads
Glass beads 3 mm
Supelco
1.04015
Glass beads 3 mm
Glass beads 2 mm
Supelco
1.04014
Glass beads 2 mm