Z261289
Stainless steel 316 syringe needle, noncoring point
gauge 20, L 6 in.
Sinonimo/i:
316 Stainless Needle, Noncoring Needle
About This Item
Codice UNSPSC:
42142600
NACRES:
NB.03
Prodotti consigliati
Descrizione
Micro-Mate Hub with 316 Stainless Steel Tubing
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Popper 9863
Lunghezza
6 in.
Gauge
20
Descrizione generale
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
Micro-Mate Hub with 316 Stainless Steel Tubing
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)