Z258709
Ace vacuum-sublimation apparatus
Complete apparatus, H × I.D. 200 mm × 60 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.76
Prodotti consigliati
Produttore/marchio commerciale
Ace Glass 802340
Alt. × diam. int.
200 mm × 60 mm
Categorie correlate
Descrizione generale
The Ace vacuum-sublimation apparatus features a greaseless O-ring sealed flange with a quick-release clamp. The top flange is ground flat, bottom mating flange has a groove for a CAPFE (fluororesin encapsulated silicone) O-ring. The complete apparatus includes an outer body, inner top, CAPFE O-ring, and quick-release clamp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z258709-1EA | 4061832363066 |