Documenti fondamentali
Z243272
ACE Squibb separatory funnel with Locktight stopper
capacity 250 mL, stopper joint: ST/NS 24/40
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Giunto
joint: ST/NS stopper24/40
Produttore/marchio commerciale
Ace Glass 723159
Capacità
250 mL
Descrizione generale
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
Note legali
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)