Key Documents
Z231592
Duran® watch glasses
O.D. 60 mm
Sinonimo/i:
213213404
About This Item
Codice UNSPSC:
41121801
NACRES:
NB.43
Produttore/marchio commerciale
DURAN®
D.E.
60 mm
Descrizione generale
With fused edges. Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance.
Note legali
DURAN is a registered trademark of DWK Life Sciences