Z200336
Squeeze-bulb pump
internal flapper valve prevents reverse flow, pkg of 1 ea
About This Item
Codice UNSPSC:
40151500
NACRES:
NB.51
Prodotti consigliati
Confezionamento
pkg of 1 ea
Categorie correlate
Descrizione generale
PE, 1 to 1 1/2 gpm. Flexible discharge tube and hose hook with tapered adapter fit 1 to 5 gal containers or cans. Internal flapper valve prevents reverse flow.
Uses: mild solvents, light oils, corrosives, dilute acid/caustics, liquid waxes, and disinfectants. Barrel: 21 in. L. Hose: 38 L × 1 in. diameter.
Uses: mild solvents, light oils, corrosives, dilute acid/caustics, liquid waxes, and disinfectants. Barrel: 21 in. L. Hose: 38 L × 1 in. diameter.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z200336-1EA | 4061832331492 |