Documenti fondamentali
Z131261
Sigma-Aldrich
Nalgene® volumetric flask
volume 250 mL, accuracy: 0.24 mL, translucent polypropylene copolymer (PPCO)
Sinonimo/i:
nalgene, plastic flask, plastic flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
long
translucent polypropylene copolymer (PPCO)
Giunto
joint threaded neck20mm
Sterilità
non-sterile
Caratteristiche
accuracy class B (ASTM E288 and ISO Standard 384)
closure type Screw cap
Confezionamento
pack of 1
Produttore/marchio commerciale
Nalgene® 4000-0250
Campo di misura del volume
250 mL, accuracy: 0.24 mL
Capacità
250 mL
Descrizione generale
Gravimetrically calibrated TC/TD at 20°C individually, then marked with a permanent color graduation line. Meet Class B accuracy requirements of ASTM E288 and ISO Standard 384. No meniscus to confuse readings. Oval marking spot. Linerless PP screw closure. Leakproof and autoclavable.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)