Z130966
Nalgene® Griffin beakers
capacity 150 mL, translucent polypropylene
Sinonimo/i:
PP beaker, nalgene PP beaker
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
translucent polypropylene
Caratteristiche
graduations
Confezionamento
pkg of 12 ea
Produttore/marchio commerciale
Nalgene 1201-0150
Capacità
150 mL
Categorie correlate
Descrizione generale
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Altre note
Browse all plastic beakers
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z130966-12EA | |
Z130966-1PAK | 4061832615653 |