Z118990
Scienceware® Frigimat® Junior dry ice maker
produces dry ice in snowflake form
About This Item
Codice UNSPSC:
41103000
NACRES:
NB.22
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F38886-0000
Descrizione generale
Produces dry ice in snowflake form. Consists of a special brass nozzle, PE cone, and a 6 × 12 in. heavy-duty cloth bag collector. The nozzle is attached directly to a cylinder (must have dip tube) of liquid CO2.
Ideal for use in remote locations, low demand or for packing or freezing samples. All components are included to connect to a standard CO2 cylinder with siphon tube to make dry ice as needed. Fittings and collection bag attach directly to the CO2 cylinder. Fits US connections.
Easy and fast dry ice flake or snow production on demand.
This device connects to CO2 gas cylinders with CGA 320 threads which is the standard for CO2 in the USA.
The Frigimate Junior Dry-Ice Maker does not fit European Gas-bottles.
Ideal for use in remote locations, low demand or for packing or freezing samples. All components are included to connect to a standard CO2 cylinder with siphon tube to make dry ice as needed. Fittings and collection bag attach directly to the CO2 cylinder. Fits US connections.
Easy and fast dry ice flake or snow production on demand.
This device connects to CO2 gas cylinders with CGA 320 threads which is the standard for CO2 in the USA.
The Frigimate Junior Dry-Ice Maker does not fit European Gas-bottles.
Note legali
Frigimat is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z118990-1EA | 4061826277225 |