Key Documents
WHA77002805
Whatman® UNIFILTER® plates
96 well, well volume 800 μL, 0.45 μm polypropylene membrane
Sinonimo/i:
Z746479, micro plate, multiwell plate, well plates
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Materiali
polypropylene
polypropylene membrane
Sterilità
non-sterile
Caratteristiche
0.45 μm polypropylene membrane
Confezionamento
pkg of 25 ea
Produttore/marchio commerciale
Whatman 7700-2805
Whatman Article No. 28420808 (US reference)
Misura
96 well
Volume pozzetto
800 μL
Dimensione pori
0.45 μm
Tipo di binding
non-treated surface
Descrizione generale
  • Design prevents well to well crosstalk
  • Large variety of well volumes economizes on expensive consumables
  • Choice of filter media allows control of flow rates and retention characteristics
  • Standard overall dimensions fit most plate readers, auto samplers, robots
Applicazioni
The UNIFILTER products are convenient, ready to use, disposable filter bottom microplates. Available in 96 well formats, the UNIFILTER microplates offer a choice of filter media that work well with the most stringent applications. UNIFILTERS can be used to process 96 samples at a time. The unique drip director design allows for analysis of the filtrate or the retentate. UNIFILTERS are available in many different variations of well formats and range from 100 μL to 2 mL, using a wide variety of filtration media.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva