Documenti fondamentali
WHA68091012
Whatman® Anotop® 10 syringe filter
0.1 μm, 50/pk
Sinonimo/i:
Whatman filter, Z677116, syringe filter, syringe filters
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Materiali
alumina based membrane
polypropylene housing
Sterilità
non-sterile
Confezionamento
pack of 50
Produttore/marchio commerciale
Whatman 6809-1012
Whatman Article No. 28420386 (US reference)
Diametro
10 mm
Volume
≤10 mL
Dimensione pori
0.1 μm
Raccordi
female Luer inlet (lock)
male Luer outlet
Descrizione generale
Whatman Anotop non sterile syringe filters from Cytiva feature an inorganic Anopore membrane in a polypropylene housing. The Anotop aluminum-based Anopore membrane is compatible with most organic solvents and aqueous samples. Aluminum-based membranes are inorganic, designed for chemical inertness and functionality in a wide range of temperatures. This syringe filter is commonly used in sample preparation for analytical instrumentation. Unlike most organic membranes, the aluminum Anopore membrane has highly defined capillary pore structures and narrow pore-size distribution. This structure enables a high level of consistency in particle retention.
Caratteristiche e vantaggi
  • Aluminum-based inorganic Anopore membrane available in three pore sizes
  • Effective with most organic solvents and aqueous samples
  • Low hold-up volume (< 20 µl for Anotop 10) for maximum sample recovery
  • Low protein binding filter minimizes protein loss
  • Capillary pore structure with narrow pore-size distribution
Compatibilità
Low protein binding. Suitable for aqueous or organic solutions.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Anotop is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)