Key Documents
WHA2105841
Whatman® lens cleaning tissue, Grade 105
W × L 100 mm × 150 mm, pkg of (25 wallets of 25 sheets)
Sinonimo/i:
Z741353, lens cleaning paper, lens paper, lens tissue
About This Item
Codice UNSPSC:
41103200
NACRES:
NB.24
Produttore/marchio commerciale
Whatman 2105-841
Whatman Article No. 28418695 (US reference)
Confezionamento
pkg of (25 wallets of 25 sheets)
Largh. × lungh.
100 mm × 150 mm
Descrizione generale
High-quality whatman lens cleaning tissue, which is chemically pure and free from silicones and other additives. Lenses and other optical surfaces made from glass, quartz or plastic can be easily scratched if you do not clean them with a very soft surface. High quality Whatman lens cleaning tissue provides the solution. The tissue is chemically pure and free from silicones and other additives. Most importantly, it can be relied on to safely remove surface moisture and grease.
Caratteristiche e vantaggi
  • Soft texture will not damage lenses or optical surfaces.
  • Chemically pure tissue is free from silicones and other additives.
  • High absorbency leads to increased safety upon removal of surface moisture and grease.
  • Very strong and leaves no fibers.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Hellmanex™ III Special Cleaning Concentrate for cuvettes
Z805939
Hellmanex III
Anti-static brush ideal for balance and lens cleaning
Z281239
Anti-static brush
Miracloth Rapid, effective filtration of homogenates
Millipore
475855
Miracloth