WHA10416294
Whatman® Nytran™ SuPerCharge (SPC) nylon blotting membranes
0.45 μm nylon blotting membranes 20 cm x 3 m roll 1/pk
Sinonimo/i:
Z670405, blotting membranes, nylon blotting membranes
About This Item
Prodotti consigliati
Materiali
Nylon membrane (Nytran SuPerCharge (SPC))
Confezionamento
pack of 1
Produttore/marchio commerciale
Whatman 10416294
Whatman Article No. 28415379 (US reference)
Largh. × lungh.
20 cm × 3 m
Dimensione pori
0.45 μm
Compatibilità
suitable for (Southern and Northern blotting; colony and plaque lifts and Dot-/Slot-Blots)
Categorie correlate
Descrizione generale
Nytran SPC membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling. With typical manufacturing techniques, increasing positive charge tends to increase background. Nytran SPC membranes are manufactured using a process that allows the combination of high positive charge with low background. Whether using radioactive or nonradioactive detection techniques, Nytran SPC consistently gives high signal with extremely low background. Whatman Nytran nylon membranes are available in two formats: Nytran N is moderately charged and Nytran SuPerCharge (SPC) has a very high positive charge.
Caratteristiche e vantaggi
- Very high positive charge.
- Improvements in the manufacturing process result in a membrane with a higher density of nylon per unit area.
- The increased charge and greater nylon density provide increased binding sites for your samples.
- Very uniform pore size and pore distribution compared to typical nylon membranes.
- Free of surface microvoids, which are common in other membranes.
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
WHA10416294 | 4061834015550 |