Key Documents
TEM-CF300CU
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
size 300 mesh, box of 25
Sinonimo/i:
CARBON FILM 300# COPPER 25PK
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 25
Misura
300 mesh
Descrizione generale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Copper grid, 300 mesh.