Documenti fondamentali
SYNS560004
Synthware 180 degree high vacuum valve
size 0-4 mm
Sinonimo/i:
Synthware S560004,
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S560004
Categorie correlate
Descrizione generale
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another. The non-rotating PTFE shaft has three O-rings, the tip O-ring making the primary seal on the precision formed tapered seat . For replacement Viton® O-ring Kit, see R7010,or R108110.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)