Documenti fondamentali
SYNS351656
Synthware high vacuum double oblique glass stopcock with hollow plug
bore 4 mm, stem O.D. 10 mm, Plug Size: 16.2/56
Sinonimo/i:
Synthware S351656
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
S351656
Foro
4 mm
D.E. stelo
10 mm
Categorie correlate
Descrizione generale
Synthware double oblique bore glass stopcock has diagonal, parallel bores that allow the connection of a single stem to both stems on the opposite side. The hollow glass plug option is precision ground with corresponding numbers on the barrel and plug. The lower vacuum chamber is evacuated via a separate 2 mm hole in the plug, which is not in the line of flow.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)