SYNP120050C
Synthware™ peptide synthesis vessel with fritted disc and PTFE stopcock
50 mL, disc O.D. 20 mm, porosity: coarse, GL 25
Sinonimo/i:
Synthware P120050C, Solid phase peptide synthesis vessel
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware P120050C
Capacità
50 mL
50 mL
Diam. est. disco
20 mm
Raccordi
GL 25
Descrizione generale
With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)