Documenti fondamentali
SYNM101005
Synthware Vacuum/Inert Gas Manifold with double bank and high vacuum valves
number of ports, 5, Hose Connections: Front-left, Rear-right
Sinonimo/i:
Synthware M101005, vacuum manifold, vacuum unit
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
Caratteristiche
number of ports 5
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware M101005
Descrizione generale
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm. With front-left and rear-right 10 mm hose connections.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)