Key Documents
SYNF901450
Synthware pear-shaped Schlenk flask with stopcock
capacity 50 mL, joint: ST/NS 14/20
Sinonimo/i:
Synthware F901450, modified Schlenk reaction flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
pear-shaped
single-neck
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F901450
Capacità
50 mL
Descrizione generale
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)