Key Documents
SYNF794500
Synthware modified evaporation flask
neck joint: ST/NS 24/40, capacity 500 mL
Sinonimo/i:
Synthware F794500, rotary evaporator flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
glass flask
round bottom flask
Giunto
joint: ST/NS neck24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F794500
Capacità
500 mL
Descrizione generale
Low profile design sits upright on bench and also provides for easy product access with a spatula. The unique flask shape is excellent for the controlled evaporation of foaming solvents such as toluene or xylene on a rotary evaporator
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)