SYNF564100
Synthware™ reaction tube with high vacuum valve
100 mL, joint: ST/NS 24/40, valve size 0-4 mm
Sinonimo/i:
Synthware F564100, Schlenk flask
About This Item
Prodotti consigliati
Materiali
colorless tube
glass tube
Giunto
joint: ST/NS24/40
Sterilità
non-sterile
Caratteristiche
graduations: no
Push fit
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Kemtech F564100
Capacità
100 mL
100 mL
Misura valvola
0-4 mm
Descrizione generale
Supplied with high vacuum valve with PTFE o-ring at tip and PTFE wiper (“umbrella”) to protect Viton® O-ring on shaft.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)