Documenti fondamentali
SYNF439100
Synthware three neck round bottom flask with angled side necks
100 mL, center joint: ST/NS 19/22, side joint: ST/NS 14/20
Sinonimo/i:
Synthware F439100, 3-neck round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
round bottom
three-neck (angled side necks)
Giunto
center joint: ST/NS19/22
side joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F439100
Capacità
100 mL
100 mL
Descrizione generale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)