SYNC414500
Synthware™ Chromatography reservoir with Rodaviss joint
capacity 500 mL, joint: ST/NS 24/40
Sinonimo/i:
Synthware C414500, glass chromatograpy column
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Prodotti consigliati
Materiali
colorless
glass
Giunto
joint: ST/NS24/40
Sterilità
non-sterile
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C414500
Capacità
500 mL
Categorie correlate
Descrizione generale
Reservoir has a top 24/40 or 45/50 Rodaviss outer joint for use with flow controller, and the listed size lower inner joint. Supplied complete with open top cap, Viton® O-ring and loosening ring.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)