Documenti fondamentali
SLW2180/08M
Pyrex® Separating funnel, pear-shape, glass key
500 mL, glass stopper, PE stopper, pack of 1 ea
Sinonimo/i:
Pyrex, SciLabware 2180/08M
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Materiali
PE stopper
PTFE key
glass stopper
pear-shaped
Giunto
joint ST/NS cone24/29
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
SciLabware 2180/08M
Diam. imbuto
107 mm
Foro rubinetto
4.0 mm
Capacità
500 mL
Descrizione generale
In liquid-liquid extractions, the two immiscible liquids separate into different layers where the liquid with the higher density collects in the lower part of the vessel and can be drained off via the stopcock.
Caratteristiche e vantaggi
  • Complies with ISO 4800 Type 2
  • Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
  • With interchangeable PTFE stopcock key
  • Supplied with both glass and polyethylene stoppers
  • The 5Litre size is supplementary to the ISO range.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)