Documenti fondamentali
SLW1523/12
Pyrex® Media-Lab Bottles, wide mouth, with polypropylene cap and pouring ring, with printed trace code
Sinonimo/i:
Pyrex, SciLabware 1523/12
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
borosilicate glass bottle
polypropylene cap
round bottle
white graduation
wide-mouth
Giunto
Joints threaded neck (GLS80)
Sterilità
non-sterile
Caratteristiche
graduations
with cap
with pouring ring
with printed trace code
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
SciLabware 1523/12
Altezza
252 mm
Diametro corpo
136 mm
Descrizione generale
  • Bottles complete with polypropylene screwcap and drip-free pouring ring
  • Manufactured from Pyrex borosilicate glass with proven temperature and chemical resistance qualities
  • GLS 80 screwthread provides increased access compared to GL 45 bottles aiding filling and pouring
  • Ideal for use with viscous liquids, pastes, powders, granules and larger solids
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White enamel graduations and marking spot
  • Autoclavable
  • Screwcaps and pouring rings can withstand temperatures up to 140 deg. C
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)