PL800GN-10
Plas-Labs neoprene glove box gloves
for 1 piece glove set, size 10 (for 8 in. ports)
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
Prodotti consigliati
Materiali
neoprene
Misura
10 (for 8 in. ports)
Caratteristiche
reusable
Confezionamento
pack of 1 pair
Produttore/marchio commerciale
Plas-Labs 800GN-10
Descrizione generale
Plas-Labs black neoprene rubber gloves for chemical resistance
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)