Documenti fondamentali
PL800GH-9-10
Plas-Labs Hypalon glove box gloves
for 1 piece glove set, size 9 (for 10 in. ports)
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
Materiali
Hypalon®
Misura
9 (for 10 in. ports)
Caratteristiche
reusable
Confezionamento
pack of 1 pair
Produttore/marchio commerciale
Plas-Labs 800GH-9-10
Descrizione generale
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Note legali
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)