Documenti fondamentali
NORTCM250
Bruker tapered PTFE caps
for NORS255001
About This Item
Codice UNSPSC:
41121800
NACRES:
SB.32
Caratteristiche
for NORS255001
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
Norell TCM250
Descrizione generale
Bruker Match tube caps are specially machined from PTFE fluoropolymer, are color coded by tube size, and correspond to the color of the tube clamp in the Match Insert Assembly, This enables quick identification of matching components. The tube cap incorporates a slightly tapered inner diameter. Initially, the cap aligns and pushes easily onto the tube, it then becomes progressively tighter as the tube reaches full depth within the cap, creating a positive seal to preserve volatile sample solutions throughout short or long term storage.
Note legali
Match is a trademark of Bruker Biospin Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)