Documenti fondamentali
MT51107675
Mettler Toledo DX series ISE half-cells electrode
Sulfide selective
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
Materiali
membrane (Solid state)
shaft (POM)
Produttore/marchio commerciale
Mettler-Toledo 51107675
Campo di misura
1.0-1.0-5
Campo di misura della temperatura
0-80 °C
pH
4-13
Descrizione generale
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your ISE half-cell, and you have a new ISE!
electrolyte for reference electrode: 1 mol/L KNO3, ISA solution: 10 mol/L NaOH
Prodotti correlati
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)