Key Documents
GF95236892
Stainless Steel - 17-7PH alloy, FeCr17Ni7
foil, 1m coil, 0.075mm thickness, annealed - condition a, precipitation hardenable
Sinonimo/i:
FE280300
About This Item
Codice UNSPSC:
11171600
NACRES:
NA.23
Forma fisica
annealed (condition a: precipitation hardenable)
foil (coil)
Produttore/marchio commerciale
Goodfellow 952-368-92
Misura × spessore
1 m × 0.075 mm