Key Documents
ESYNTH002
Sigma-Aldrich
SynLectro 200 mL starter kit
Sinonimo/i:
SynLectro 200 mL Beginning Set
About This Item
Codice UNSPSC:
81141502
NACRES:
NB.61
Descrizione generale
SynLectro 200 mL Starter Kit is the perfect way to get your electrochemical reaction started.
This kit contains:
  • 200 mL glass cell
  • PTFE stopper and sleeve
  • 2 electrode holders
  • 2 graphite electrodes
  • stainless steel electrode
  • nickel electrode
  • lead electrode
  • zinc electrode
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Note legali
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)