Documenti fondamentali
DWKW242821-A
Wheaton® Dropping Bottle
natural bottle, capacity 3 mL, case of 1000
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
natural bottle
Confezionamento
pkg of 1 packs
case of 1000
Produttore/marchio commerciale
WHEATON® W242821-A
Capacità
3 mL
Applicazioni
Opthalmic, IVD Kits, Other
Caratteristiche e vantaggi
  • Bottle made of natural or white LDPE (Low Density Polyethylene
  • White LDPE bottle helps protect UV light sensitive samples
  • Dropper tip caps and tips sold separately
  • Packaged in cases of 100 or 1000
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
DWKW242821-A-1PK