DWK14395-1000
KIMBLE® KIMAX® Media bottle
with cap, round transparent borosilicate glass bottle, bottle capacity (1,000 mL), non-sterile
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
blue polypropylene cap
round transparent borosilicate glass bottle
Sterilità
non-sterile
Caratteristiche
with cap
Confezionamento
case of 10 ea
Produttore/marchio commerciale
Kimble® 14395-1000
Capacità bottiglia
(1,000 mL)
Raccordi
GL 45 neck
GL 45
Descrizione generale
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
- Enhanced graduations and marking spot made with chemically resistant white enamel paint
- 30 mm ID opening
- Autoclavable
- Supplied without caps or with linerless GL 45 screw thread caps
- Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)