Documenti fondamentali
DWK1165888
Duran® GLS 80 caps and accessories
GLS80, high temp, white, PSU, 5EA
Sinonimo/i:
1165888
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
white
Confezionamento
case of 5 ea
Produttore/marchio commerciale
DURAN® 1165888
Diam. tappo
(88.5 mm)
Descrizione generale
PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. The matching PTFE pouring ring permitts drip-free pouring and clean, safe working
Caratteristiche e vantaggi
• Manufactured from Polysulfone (PSU), PTFE coated, platinum-cured silicone• PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components• Screw cap seal with PTFE coated, platinum-cured silicone for efficient sealing• The matching PTFE pouring ring permits drip-free pouring and clean, safe working• Suitable for autoclaving / steam sterilisation and for use up to 180 °C
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)