CLS500050
Pyrex® narrow-mouth Erlenmeyer flask with joint
capacity 50 mL
Sinonimo/i:
Pyrex® wide-mouth Erlenmeyer flask with joint, conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5000
About This Item
Prodotti consigliati
Materiali
borosilicate glass
bottom (baffled)
glass cap
narrow-mouth
Giunto
joint ST/NS19/38
Sterilità
non-sterile
Caratteristiche
cap
closure type Standard Taper joint
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 5000-50
Capacità
50 mL
Descrizione generale
- Pyrex® Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
- Flasks have durable white enamel graduations to show approximate capacity.
- Large marking spot for labeling.
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
C85409-25G | 4061833543115 |
C85409-5G | 4061833522417 |
CLS500050-1EA | 4061825882864 |