Documenti fondamentali
CLS401901
Corning® Modified Impeller Assembly (for use with probes) for Magnetically-Driven Proculture® Spinner Flask
fits flask, 36 L
Sinonimo/i:
equipment replacement accessory
About This Item
Codice UNSPSC:
41122104
NACRES:
NB.43
Materiali
stainless steel shaft
Sterilità
non-sterile
Caratteristiche
fits flask 36 L
Produttore/marchio commerciale
Corning 401901
Descrizione generale
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 36L Proculture Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
Note legali
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)