Documenti fondamentali
CLS3920M
Pyrex® continuous liquid/liquid extractor
complete
Sinonimo/i:
liquid liquid extraction, liquid liquid extractor, liquid-liquid extraction, liquid-liquid extractor, Corning® 3920M
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.43
Produttore/marchio commerciale
Corning 3920-M
Descrizione generale
Pyrex brand continuous liquid/liquid extractor is designed to meet E.P.A. test method requirements outlined by SW-846, RCRA, and CLP. For use with solvents of heavier specific gravity than sample liquid being extracted. The sidearm is sealed to the extractor body just below the 45/50 ground joint. The sidearm is sealed higher on the body to reduce the chance of water spilling over while draining remaining solvent from extractor body at the end of the extraction. Extraction can be performed outside the hood. Comes complete with Pyrex brand Allihn condenser and 500mL Pyrex brand round bottom boiling flask. As dictated by safe laboratory practices, we recommend that some type of ventilation system be used above or near the units in case of a drop in condenser water temperature or loss of condenser cold water supply.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)