Documenti fondamentali
BR91210
BRAND® glass beaker with spout, low form
volume 5 mL, without graduation
Sinonimo/i:
Beakers, Glass
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Materiali
borosilicate glass 3.3
Caratteristiche
without graduation
Produttore/marchio commerciale
BRAND 91210
Volume
5 mL
Descrizione generale
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer. (no graduations)
Altre note
Browse all glass beakers
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
BR91210-10EA4061831861624