BR90654
BRAND® glass beaker with spout, tall form
volume 1000 mL, with graduation
Sinonimo/i:
Beakers, Glass
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass 3.3
Caratteristiche
with graduation
Produttore/marchio commerciale
BRAND 90654
Volume
1000 mL
Categorie correlate
Descrizione generale
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Altre note
Browse all glass beakers
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR90654-10EA | 4061831867794 |