BR90256
BRAND® ETFE beaker with spout, low form
volume 600 mL, with graduation
Sinonimo/i:
Beakers, Plastics
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
ETFE
Caratteristiche
with graduation
Produttore/marchio commerciale
BRAND 90256
Volume
600 mL
Categorie correlate
Descrizione generale
Made from high-quality ETFE (ethylene-tetrafluoroethylene), this translucent beaker offers excellent chemical resistance, making it ideal for handling various substances in laboratory settings. The low form provides stability and ease of use, allowing for efficient mixing and pouring of liquids. Clearly marked graduations facilitate accurate measurement of liquids, ensuring precision in experiments and formulations. The beaded rim enhances strength and provides a comfortable grip, making handling safer and more efficient.
Altre note
Browse all plastic beakers
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR90256-1EA | 4061831861495 |