BR88635
BRAND® stopcock one-way with nozzles
for tube I.D. 8 mm
Sinonimo/i:
PTFE, Stopcocks
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Prodotti consigliati
Materiali
PTFE
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 88635
Diam. int.
4 mm
Categorie correlate
Descrizione generale
The BRAND® one-way stopcock with nozzles is constructed from high-quality PTFE, offering outstanding chemical resistance for various laboratory applications. This stopcock features a valve lever made of polypropylene (PP) for easy operation and control. It is equipped with integrated hose nozzles, facilitating efficient fluid transfer and connection to hoses. The robust design ensures reliable performance, making it suitable for handling aggressive chemicals and solvents.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR88635-1EA | 4061831861358 |