BR81265
BRAND® spare key for automatic burettes Pellet pattern
PTFE key
Sinonimo/i:
Accessories, Burette stopcock, Burettes, Compact Burettes, Stopcocks
About This Item
Codice UNSPSC:
41121808
NACRES:
NB.43
Prodotti consigliati
Materiali
PTFE key
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 81265
Descrizione generale
The BRAND® spare key for automatic burettes features a pellet pattern design and is made from durable PTFE, ensuring chemical resistance and longevity. This spare key is equipped with a retention device, providing secure attachment and preventing accidental loss during use. It is specifically designed to enhance the functionality of automatic burettes, allowing for smooth and precise liquid dispensing.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)