Documenti fondamentali
BR782153
BRAND® lids for microplates BRAND®plates
plate size 1536 wells, without condensation rings
Sinonimo/i:
Accessories, BRANDplates®, Lids, Microplates
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
polystyrene
Confezionamento
pack of 50 ea
Produttore/marchio commerciale
BRAND 782153
Dimens. piastra
1536 wells
Misura
1536 wells
Compatibilità
suitable for (bacteriological assays)
Descrizione generale
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)