BR707537
BRAND® dispenser cylinder with valve block for Titrette®
for 50 mL from serial-number 01K
Sinonimo/i:
Accessories, Bottle-top Burette, Burettes, Titrette®
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 707537
Descrizione generale
Structural change from serial-number 01K (January 2012).The assembly of the titrating tube to the valve block has been changed. When ordering spare parts, the serial number must be accounted for.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)