Documenti fondamentali
BR704541
BRAND® pump assembly seripettor®
size 2 mL
Sinonimo/i:
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
Codice UNSPSC:
41121516
NACRES:
NB.01
Materiali
polycarbonate
stainless steel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 704541
Misura
2 mL
Categorie correlate
Descrizione generale
The BRAND® Pump Assembly for Seripettor® features a high-quality actuating unit designed for precise liquid dispensing. Constructed from durable polycarbonate (PC), this assembly ensures longevity and reliability in laboratory applications. The stainless steel lifting spring enhances performance, providing smooth and efficient operation. This pump assembly is an essential component of the Seripettor® system, making it ideal for a variety of liquid handling tasks in research and analytical settings.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)