BR51467
BRAND® ground socket
PTFE, joint: 29/32, grip collar and sealing rings
Sinonimo/i:
Ground sockets, PTFE
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.43
Prodotti consigliati
Materiali
PTFE
Giunto
joint: 29/32
Caratteristiche
grip collar and sealing rings
Produttore/marchio commerciale
BRAND 51467
Categorie correlate
Descrizione generale
PTFE. Greaseless seal will not contaminate sample. Outstanding chemical resistance at operating temperatures from -200 to +260 °C. Sturdy design for permanent use. With grip collar and sealing rings for excellent tightness. Low leakage rate: Q < 1 · 10-4 mbar · l · s-1.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR51467-1EA | 4061831857375 |