Documenti fondamentali
BR4720420
BRAND® seripettor® pro bottle-top dispenser
volume range 0.2-2 mL
Sinonimo/i:
Bottle-top Dispenser, Dispenser, seripettor®
About This Item
Codice UNSPSC:
41121516
NACRES:
NB.01
Caratteristiche
incremental 0.04 mL
Produttore/marchio commerciale
BRAND 4720420
Intervallo di volume
0.2-2 mL
Raccordi
(GL 45/32)
(GL 45/38)
(GL 45/S 40)
Descrizione generale
The seripettors are innovative bottle-top dispensers from BRAND with a special design. A lifting spring provides automatic filling. When the piston seal is worn out, the entire dispensing unit can be exchanges by the user rapidly and problem-free, without tools - Eliminating expensive repairs. Seripettor pro developed specifically for dispensing chemicals.
Not compatible with hydrofluoric acid, or other chemicals that attack polypropylene, PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, borosilicate glass and carbon disulfide
Caratteristiche e vantaggi
  • Low-cost replaceable dispensing cartridge with wiping seal - one spare dispensing cartridge included
  • Semi-automatic dispensing - The spring loaded piston refills the cylinder after dispensing
  • Volume setting - Scalloped track allows for quick and exact setting of desired volume
  • Disassembles without tools - All parts can be replaced quickly without tools
  • The innovate design makes cleaning and maintenance easy
  • Supplied with discharge tube, filling tube, spare dispensing cartridge and adapters, PP (GL 45/32 and GL 45/540)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)